Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020624.1 | NP_001018460.1 | 2 - 3 | 232 | 124 | |
GenBank mRNA | BC095370.1 | - | 2 - 3 | 232 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |