Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pygma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020628.1 | NP_001018464.1 | 17 - 18 | 2244 | 89 | |
GenBank mRNA | BC095379.1 | - | 17 - 18 | 2244 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |