Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024951.2 | NP_001020122.2 | 1 - 2 | 331 | 80 | |
XM_017356442.2 | XP_017211931.1 | 1 - 2 | 336 | 80 | ||
GenBank mRNA | AF265345.1 | - | 1 - 2 | 296 | 80 | |
BC071299.1 | - | 1 - 2 | 321 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |