Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024951.2 | NP_001020122.2 | 5 - 6 | 627 | 87 | |
XM_017356442.2 | XP_017211931.1 | 6 - 7 | 671 | 87 | ||
GenBank mRNA | AF265345.1 | - | 5 - 6 | 87 | ||
BC071299.1 | - | 5 - 6 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |