Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025469.1 | NP_001020640.1 | 1 - 2 | 124 | 87 | |
GenBank mRNA | BC095279.1 | - | 1 - 2 | 124 | 87 | |
BC171513.1 | - | 1 - 2 | 113 | 87 | ||
BC171519.1 | - | 1 - 2 | 113 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |