Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025469.1 | NP_001020640.1 | 2 - 3 | 186 | 85 | |
GenBank mRNA | BC095279.1 | - | 2 - 3 | 186 | 85 | |
BC171513.1 | - | 2 - 3 | 175 | 85 | ||
BC171519.1 | - | 2 - 3 | 175 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |