Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025510.1 | NP_001020681.1 | 1 - 2 | 376 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |