Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025556.1 | NP_001020727.1 | 1 - 2 | 360 | 142 | |
GenBank mRNA | BC096889.1 | - | 1 - 2 | 360 | 142 | |
BC164179.1 | - | 1 - 2 | 374 | 142 | ||
EF014961.1 | - | 1 - 2 | 355 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |