Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrm5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030160.2 | NP_001025331.1 | 1 | 1148 | 68 | |
GenBank mRNA | HM038481.1 | - | 1 | 1015 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |