Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030220.1 | NP_001025391.1 | 14 - 15 | 1473 | 73 | |
XM_017352511.2 | XP_017208000.1 | 13 - 14 | 2073 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Probe spans exons |