Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040249.2 | NP_001035339.2 | 9 - 10 | 1401 | 81 | |
XM_021479195.1 | XP_021334870.1 | 9 - 10 | 1343 | 81 | ||
GenBank mRNA | BC115303.1 | - | 8 - 9 | 1116 | 81 | |
BC124075.1 | - | 9 - 10 | 1401 | 81 | ||
BC164676.1 | - | 8 - 9 | 986 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |