Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pglyrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045166.1 | NP_001038631.1 | 3 - 4 | 1055 | 90 | |
XM_021476909.1 | XP_021332584.1 | 3 - 4 | 1333 | 90 | ||
GenBank mRNA | DQ447202.1 | - | 3 - 4 | 1055 | 90 | |
DQ482738.1 | - | 3 - 4 | 992 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |