Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lect2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048055.1 | NP_001041520.1 | 1 - 2 | 155 | 82 | |
GenBank mRNA | BC162786.1 | - | 1 - 2 | 92 | 82 | |
BC162799.1 | - | 1 - 2 | 92 | 82 | ||
DQ372604.1 | - | 1 - 2 | 155 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |