Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110460.1 | NP_001103930.1 | 2 - 3 | 490 | 78 | |
XM_005155544.3 | XP_005155601.1 | 2 - 3 | 513 | 78 | ||
XM_005155545.3 | XP_005155602.1 | 2 - 3 | 513 | 78 | ||
XM_005155546.3 | XP_005155603.1 | 2 - 3 | 513 | 78 | ||
XM_005155547.3 | XP_005155604.1 | 2 - 3 | 513 | 78 | ||
XM_021479184.1 | XP_021334859.1 | 2 - 3 | 513 | 78 | ||
GenBank mRNA | BC139674.1 | - | 2 - 3 | 490 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |