Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110460.1 | NP_001103930.1 | 6 - 7 | 1041 | 91 | |
XM_005155544.3 | XP_005155601.1 | 6 - 7 | 1064 | 91 | ||
XM_005155545.3 | XP_005155602.1 | 6 - 7 | 1064 | 91 | ||
GenBank mRNA | BC139674.1 | - | 6 - 7 | 1041 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |