Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: parvab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110471.1 | NP_001103941.1 | 7 - 8 | 781 | 65 | |
GenBank mRNA | BC134924.1 | - | 7 - 8 | 781 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |