Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: parvab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110471.1 | NP_001103941.1 | 8 - 9 | 806 | 62 | |
GenBank mRNA | BC134924.1 | - | 8 - 9 | 806 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |