Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dharma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130979.3 | NP_571054.1 | 1 - 2 | 360 | 105 | |
GenBank mRNA | AB010103.2 | - | 1 - 2 | 366 | 105 | |
AF098202.1 | - | 1 - 2 | 315 | 105 | ||
BC122095.1 | - | 1 - 2 | 362 | 105 | ||
BC165258.1 | - | 1 - 2 | 325 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |