Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dr03099385_m1
See other NCAM1B GE Assays ›
Gene Symbol:
ncam1b
Gene Name:
neural cell adhesion molecule 1b
Gene Aliases:
ncam3, zPCAM
Chromosome Location:
Chr.15: 18367307 - 18574863 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03099385_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ncam1b
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_131831.1
NP_571906.1
13 - 14
1890
63
XM_005157405.4
XP_005157462.1
14 - 15
2061
63
XM_005157406.4
XP_005157463.1
13 - 14
2037
63
XM_005157407.4
XP_005157464.1
14 - 15
2061
63
XM_005157408.4
XP_005157465.1
14 - 15
2061
63
XM_005157409.4
XP_005157466.1
12 - 13
2007
63
XM_005157410.4
XP_005157467.1
13 - 14
2036
63
XM_005157411.4
XP_005157468.1
12 - 13
2006
63
XM_009291549.3
XP_009289824.1
14 - 15
2061
63
XM_009291550.3
XP_009289825.1
14 - 15
2061
63
XM_009291551.3
XP_009289826.1
14 - 15
2061
63
XM_009291552.3
XP_009289827.1
13 - 14
2031
63
XM_009291553.3
XP_009289828.1
14 - 15
2061
63
XM_009291554.3
XP_009289829.1
14 - 15
2061
63
XM_009291556.3
XP_009289831.1
14 - 15
2061
63
XM_009291557.3
XP_009289832.1
14 - 15
2061
63
XM_009291558.3
XP_009289833.1
14 - 15
2061
63
XM_009291559.3
XP_009289834.1
12 - 13
2007
63
XM_017359195.2
XP_017214684.1
13 - 14
2037
63
GenBank mRNA
AY029401.1
-
13 - 14
1890
63
BC162073.1
-
13 - 14
1790
63
CU458747.1
-
13 - 14
1680
63
CU458882.1
-
14 - 15
1704
63
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Mammalian Gene Collection
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
cell adhesion
axon guidance
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0