Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199333.1 | NP_955365.1 | 10 - 11 | 1549 | 113 | |
XM_009293364.2 | XP_009291639.1 | 10 - 11 | 1662 | 113 | ||
GenBank mRNA | BC045421.1 | - | 10 - 11 | 1549 | 113 | |
BC067143.1 | - | 9 - 10 | 1530 | 113 | ||
BC164692.1 | - | 9 - 10 | 1528 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |