Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zbtb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200160.1 | NP_956454.1 | 2 - 3 | 324 | 72 | |
XM_005158599.4 | XP_005158656.1 | 2 - 3 | 594 | 72 | ||
GenBank mRNA | BC045363.1 | - | 2 - 3 | 324 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |