Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200257.1 | NP_956551.1 | 4 - 5 | 1759 | 75 | |
GenBank mRNA | BC048060.1 | - | 4 - 5 | 1759 | 75 | |
BC164039.1 | - | 3 - 4 | 1675 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |