Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cxxc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200599.1 | NP_956893.1 | 8 - 9 | 1116 | 75 | |
GenBank mRNA | BC056775.1 | - | 8 - 9 | 1116 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |