Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcmtl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200768.1 | NP_957062.1 | 6 - 7 | 536 | 64 | |
XM_005158766.4 | XP_005158823.1 | 7 - 8 | 643 | 64 | ||
XM_005158767.4 | XP_005158824.1 | 6 - 7 | 552 | 64 | ||
XM_009293125.3 | XP_009291400.1 | 7 - 8 | 643 | 64 | ||
GenBank mRNA | BC059599.1 | - | 6 - 7 | 536 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |