Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gipc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200930.1 | NP_957224.1 | 3 - 4 | 852 | 88 | |
GenBank mRNA | AY989814.1 | - | 3 - 4 | 632 | 88 | |
BC045953.1 | - | 3 - 4 | 852 | 88 | ||
BC064297.1 | - | 3 - 4 | 822 | 88 | ||
BC155746.1 | - | 3 - 4 | 637 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |