Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201139.2 | NP_957433.2 | 7 - 8 | 1008 | 81 | |
GenBank mRNA | BC044552.1 | - | 7 - 8 | 1004 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |