Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205677.1 | NP_991240.1 | 4 - 5 | 670 | 67 | |
GenBank mRNA | BC065467.1 | - | 4 - 5 | 670 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |