Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205698.1 | NP_991261.1 | 2 - 3 | 565 | 85 | |
XM_021466666.1 | XP_021322341.1 | 2 - 3 | 661 | 85 | ||
GenBank mRNA | AY845128.1 | - | 2 - 3 | 573 | 85 | |
BC053310.1 | - | 3 - 4 | 516 | 85 | ||
BC065966.1 | - | 2 - 3 | 565 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |