Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212657.1 | NP_997822.1 | 1 - 2 | 213 | 65 | |
GenBank mRNA | BC048894.1 | - | 1 - 2 | 213 | 65 | |
BC164197.1 | - | 1 - 2 | 67 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |