Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssrp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212802.2 | NP_997967.2 | 9 - 10 | 1166 | 73 | |
GenBank mRNA | BC056311.1 | - | 9 - 10 | 1072 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |