Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213495.1 | NP_998660.1 | 19 - 20 | 2299 | 101 | |
XM_021470126.1 | XP_021325801.1 | 19 - 20 | 2386 | 101 | ||
GenBank mRNA | BC048370.1 | - | 19 - 20 | 2299 | 101 | |
BC165368.1 | - | 18 - 19 | 2065 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |