Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtpbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002454.1 | NP_001002454.1 | 4 - 5 | 535 | 72 | |
GenBank mRNA | BC076017.1 | - | 4 - 5 | 535 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |