Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003651.1 | NP_001003651.1 | 4 - 5 | 277 | 78 | |
XM_021470141.1 | XP_021325816.1 | 4 - 5 | 293 | 78 | ||
GenBank mRNA | BC076430.1 | - | 4 - 5 | 277 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |