Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003869.1 | NP_001003869.1 | 2 - 3 | 440 | 83 | |
GenBank mRNA | AY648791.1 | - | 2 - 3 | 440 | 83 | |
BC093104.1 | - | 2 - 3 | 388 | 83 | ||
BC115114.1 | - | 2 - 3 | 391 | 83 | ||
BC121728.1 | - | 2 - 3 | 359 | 83 | ||
BC129281.1 | - | 2 - 3 | 389 | 83 | ||
BC164482.1 | - | 1 - 2 | 276 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |