Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005578.2 | NP_001005578.2 | 1 - 2 | 64 | 85 | |
GenBank mRNA | AY570235.1 | - | 1 - 2 | 63 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Danio rerio Probe spans exons |