Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012482.1 | NP_001012500.1 | 21 - 22 | 3228 | 89 | |
XM_009307525.2 | XP_009305800.1 | 21 - 22 | 3240 | 89 | ||
XM_009307527.2 | XP_009305802.1 | 20 - 21 | 3225 | 89 | ||
XM_017358709.2 | XP_017214198.1 | 21 - 22 | 3240 | 89 | ||
XM_017358710.2 | XP_017214199.1 | 21 - 22 | 3240 | 89 | ||
XM_017358711.2 | XP_017214200.1 | 21 - 22 | 3240 | 89 | ||
XM_017358712.2 | XP_017214201.1 | 21 - 22 | 3240 | 89 | ||
XM_017358713.2 | XP_017214202.1 | 21 - 22 | 3240 | 89 | ||
XM_017358714.2 | XP_017214203.1 | 21 - 22 | 3240 | 89 | ||
GenBank mRNA | AY772390.1 | - | 21 - 22 | 3228 | 89 | |
KY432405.1 | - | 21 - 22 | 3227 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |