Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013269.3 | NP_001013287.2 | 3 - 4 | 885 | 76 | |
GenBank mRNA | BC107955.1 | - | 3 - 4 | 889 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |