Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013479.1 | NP_001013497.1 | 6 - 7 | 501 | 129 | |
XM_021480064.1 | XP_021335739.1 | 6 - 7 | 354 | 129 | ||
GenBank mRNA | BC091449.1 | - | 6 - 7 | 501 | 129 | |
BC164838.1 | - | 6 - 7 | 451 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |