Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmnat1-rbp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017629.2 | NP_001017629.1 | 2 - 3 | 151 | 65 | |
GenBank mRNA | BC092821.1 | - | 2 - 3 | 153 | 65 | |
BC165446.1 | - | 1 - 2 | 143 | 65 | ||
CO804002.1 | - | 2 - 3 | 161 | 65 | ||
CT621800.2 | - | 1 - 2 | 134 | 65 |
Gene Symbol: nmnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328328.1 | NP_001315257.1 | 2 - 3 | 166 | 65 | |
GenBank mRNA | CK016190.1 | - | 1 - 2 | 121 | 65 | |
EH495187.1 | - | 2 - 3 | 192 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |