Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017883.1 | NP_001017883.1 | 2 - 3 | 165 | 99 | |
GenBank mRNA | BC092965.1 | - | 2 - 3 | 165 | 99 | |
BC164333.1 | - | 2 - 3 | 183 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |