Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017883.1 | NP_001017883.1 | 4 - 5 | 393 | 109 | |
GenBank mRNA | BC092965.1 | - | 4 - 5 | 393 | 109 | |
BC164333.1 | - | 4 - 5 | 411 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |