Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pudp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020615.1 | NP_001018451.1 | 3 - 4 | 526 | 75 | |
GenBank mRNA | BC095345.1 | - | 3 - 4 | 526 | 75 | |
BC165028.1 | - | 3 - 4 | 496 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |