Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amacr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020626.1 | NP_001018462.1 | 4 - 5 | 646 | 68 | |
GenBank mRNA | BC095377.1 | - | 4 - 5 | 646 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |