Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v0a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020666.1 | NP_001018502.1 | 20 - 21 | 2368 | 65 | |
XM_009297243.2 | XP_009295518.1 | 20 - 21 | 2492 | 65 | ||
GenBank mRNA | BC095691.1 | - | 20 - 21 | 2368 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |