Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024665.1 | NP_001019836.1 | 5 - 6 | 1115 | 71 | |
XM_005160883.4 | XP_005160940.1 | 5 - 6 | 1083 | 71 | ||
GenBank mRNA | DQ060246.1 | - | 5 - 6 | 1115 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |