Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trnau1apb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029966.2 | NP_001025137.2 | 5 - 6 | 519 | 127 | |
XM_021481362.1 | XP_021337037.1 | 5 - 6 | 540 | 127 | ||
GenBank mRNA | BC098884.1 | - | 5 - 6 | 519 | 127 | |
CV489035.1 | - | 5 - 6 | 529 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |