Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tas1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039525.2 | NP_001034614.2 | 5 - 6 | 1543 | 72 | |
GenBank mRNA | AB200899.2 | - | 5 - 6 | 1543 | 72 | |
AB289804.1 | - | 5 - 6 | 1387 | 72 | ||
BC162412.1 | - | 5 - 6 | 1569 | 72 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |