Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040096.2 | NP_001035185.1 | 2 - 3 | 310 | 87 | |
GenBank mRNA | BC125893.1 | - | 2 - 3 | 310 | 87 | |
BC164327.1 | - | 2 - 3 | 286 | 87 | ||
DQ395301.1 | - | 2 - 3 | 327 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |