Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pip4p2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045112.3 | NP_001038577.1 | 6 - 7 | 685 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Danio rerio Probe spans exons |