Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048064.2 | NP_001041529.2 | 53 - 54 | 5746 | 131 | |
XM_017352790.2 | XP_017208279.1 | 52 - 53 | 6021 | 131 | ||
XM_017352791.2 | XP_017208280.1 | 51 - 52 | 5994 | 131 | ||
XM_017352793.2 | XP_017208282.1 | 52 - 53 | 6021 | 131 | ||
XM_017352794.2 | XP_017208283.1 | 51 - 52 | 5952 | 131 | ||
XM_017352796.2 | XP_017208285.1 | 48 - 49 | 5385 | 131 | ||
XM_021468860.1 | XP_021324535.1 | 51 - 52 | 5841 | 131 | ||
XM_021468861.1 | XP_021324536.1 | 50 - 51 | 5766 | 131 | ||
XM_021468862.1 | XP_021324537.1 | 46 - 47 | 5145 | 131 | ||
GenBank mRNA | DQ523563.1 | - | 53 - 54 | 5848 | 173 | |
MF118597.1 | - | 5 - 6 | 462 | 173 | ||
MF118598.1 | - | 5 - 6 | 405 | 173 | ||
MF118599.1 | - | 5 - 6 | 462 | 173 | ||
MF118600.1 | - | 5 - 6 | 462 | 173 | ||
MF118601.1 | - | 5 - 6 | 462 | 173 | ||
MF118602.1 | - | 5 - 6 | 405 | 173 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |